4-Ethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A149180-25G A149180-5G A149180-100G
77286-062EA 13 USD
77286-062 77286-064 77615-202
4-Ethoxybenzaldehyde 98%
4-Ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00003388
CAS Number: 10031-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR