2-Methylbenzothiazol-6-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylbenzo[d]thiazol-6-ol

A149164-5G A149164-1G
77259-150EA 213.21 USD
77259-150 77567-110
2-Methylbenzothiazol-6-ol 97%
2-Methylbenzothiazol-6-ol

Precaución: For research use only.

Formula: C₈H₇NOS
MW: 165.21 g/mol
MDL Number: MFCD00227234
CAS Number: 68867-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR