1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Butyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)
A149153-100G
A149153-1G
A149153-25G
A149153-500G
A149153-5G
77235-350EA
64.89
USD
77235-350
77235-352
77235-354
77235-356
77235-358
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate 99%
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate
Precaución: for Research Use Only
Formula:
C₉H₁₇F₆N₂P MW: 298.21 g/mol |
MDL Number:
MFCD03790877 CAS Number: 227617-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...