Methyl-3-aminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, β-Alanine methyl ester hydrochloride, 3-Aminopropionic acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl

A149036-100G A149036-10G A149036-25G A149036-500G A149036-5G
77272-904EA 36.05 USD
77272-904 77272-906 77272-908 77272-910 77272-912
Methyl-3-aminopropanoate hydrochloride 97%
Methyl-3-aminopropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉NO₂·HCl
MW: 139.6 g/mol
MDL Number: MFCD00039060
CAS Number: 3196-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR