3,5-Difluoro-2-nitrophenol 97%

Supplier: Ambeed
Danger

A148649-10G A148649-1G A148649-250MG A148649-25G A148649-5G
77272-888EA 224.54 USD
77272-888 77272-890 77272-892 77272-894 77272-896
3,5-Difluoro-2-nitrophenol 97%
3,5-Difluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MW: 175.0900 g/mol
MDL Number: MFCD08458165
CAS Number: 151414-46-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR