2'-Fluoro-2'-deoxycytidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 2'-Deoxy-2'-fluorocytidine

A148637-1G A148637-250MG A148637-5G
77272-882EA 41.2 USD
77272-882 77272-884 77272-886
2'-Fluoro-2'-deoxycytidine 95%
2'-Fluoro-2'-deoxycytidine

Precaución: for Research Use Only

Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
MDL Number: MFCD00057445
CAS Number: 10212-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR