1-Amino-2,2,2-trifluoroethane hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoroethanamine hydrochloride, 2,2,2-Trifluoroethylamine hydrochloride

A148633-100G A148633-10G A148633-1G A148633-25G A148633-500G A148633-5G
77247-178EA 37.08 USD
77247-178 77247-180 77247-182 77247-184 77247-186 77247-188
1-Amino-2,2,2-trifluoroethane hydrochloride 98%
1-Amino-2,2,2-trifluoroethane hydrochloride

Precaución: for Research Use Only

Formula: C₂H₅ClF₃N
MW: 135.52 g/mol
MDL Number: MFCD00012875
CAS Number: 373-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR