3-Phenylazetidine hydrochloride 97%

Supplier: Ambeed
Warning

A148547-250MG A148547-100MG
77286-044EA 151 USD
77286-044 77549-836
3-Phenylazetidine hydrochloride 97%
3-Phenylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁N·ClH
MDL Number: MFCD08703190
CAS Number: 7606-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR