(S)-2-Amino-2-(2-chlorophenyl)ethanol hydrochloride 97%

Supplier: Ambeed
Warning

A148428-100MG A148428-1G A148428-250MG
77247-168EA 58.71 USD
77247-168 77247-170 77247-172
(S)-2-Amino-2-(2-chlorophenyl)ethanol hydrochloride 97%
(S)-2-Amino-2-(2-chlorophenyl)ethanol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂NO
MW: 208.09 g/mol
MDL Number: MFCD12758099
CAS Number: 1269773-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR