SR-12813 98%

Supplier: Ambeed
Warning

Synonyms: SR12813, Tetraethyl (2-(3,5-di-tert-butyl-4-hydroxyphenyl)ethene-1,1-diyl)bis(phosphonate)

A148389-10MG A148389-1MG A148389-250MG A148389-25MG A148389-5MG
77259-122EA 33 USD
77259-122 77259-124 77259-126 77259-128 77259-130
SR-12813 98%
SR-12813

Precaución: for Research Use Only

Formula: C₂₄H₄₂O₇P₂
MDL Number: MFCD00888473
CAS Number: 126411-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR