3-Aminopyridine-2(1H)-thione 97%

Supplier: Ambeed
Warning

A148358-100MG A148358-250MG A148358-5G
77235-314EA 12 USD
77235-314 77235-316 77235-318
3-Aminopyridine-2(1H)-thione 97%
3-Aminopyridine-2(1H)-thione

Precaución: for Research Use Only

Formula: C₅H₆N₂S
MDL Number: MFCD09757485
CAS Number: 38240-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR