Silver(I) dibenzyl phosphate 97%

Supplier: Ambeed
Warning

A148096-100MG A148096-250MG
77247-162EA 22.66 USD
77247-162 77247-164
Silver(I) dibenzyl phosphate 97%
Silver(I) dibenzyl phosphate

Precaución: for Research Use Only

Formula: C₁₄H₁₄AgO₄P
MW: 385.1 g/mol
MDL Number: MFCD00067264
CAS Number: 50651-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR