trans-4-((9-((S)-tetrahydrofuran-3-yl)-8-((2,4,6-trifluorophenyl)amino)-9H-purin-2-yl)amino)cyclohexanol 98%

Supplier: Ambeed
Warning

A148034-10MG A148034-1MG A148034-25MG A148034-2MG A148034-5MG
77235-294EA 52 USD
77235-294 77235-296 77235-298 77235-300 77235-302
trans-4-((9-((S)-tetrahydrofuran-3-yl)-8-((2,4,6-trifluorophenyl)amino)-9H-purin-2-yl)amino)cyclohexanol 98%
trans-4-((9-((S)-tetrahydrofuran-3-yl)-8-((2,4,6-trifluorophenyl)amino)-9H-purin-2-yl)amino)cyclohexanol

Precaución: for Research Use Only

Formula: C₂₁H₂₃F₃N₆O₂
MDL Number: MFCD23160040
CAS Number: 899805-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR