4-Bromo-2-fluorobenzenesulfonamide 95%

Supplier: Ambeed
Warning

A147995-1G A147995-5G
77286-020EA 20 USD
77286-020 77286-022
4-Bromo-2-fluorobenzenesulfonamide 95%
4-Bromo-2-fluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₅BrFNO₂S
MDL Number: MFCD03094291
CAS Number: 214210-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR