1-(5-(2-Fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)-N-methylmethanamine 98%

Supplier: Ambeed
Warning

A147862-1MG A147862-250MG
77235-284EA 5 USD
77235-284 77746-034
1-(5-(2-Fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)-N-methylmethanamine 98%
1-(5-(2-Fluorophenyl)-1-(pyridin-3-ylsulfonyl)-1H-pyrrol-3-yl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₁₇H₁₆FN₃O₂S
MDL Number: MFCD27987913
CAS Number: 881681-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR