2,5,6-Trichloropyrimidin-4-amine 98%

Supplier: Ambeed
Warning

A147850-250MG A147850-5G A147850-1G
77286-018EA 65 USD
77286-018 77615-180 77748-029
2,5,6-Trichloropyrimidin-4-amine 98%
2,5,6-Trichloropyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₄H₂Cl₃N₃
MDL Number: MFCD09744035
CAS Number: 28969-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR