1-Methylguanidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-Methylguanidine monohydrochloride, Methylguanidinium chloride

A147840-100G A147840-25G A147840-5G
77235-278EA 210.12 USD
77235-278 77235-280 77235-282
1-Methylguanidine hydrochloride 98%
1-Methylguanidine hydrochloride

Precaución: for Research Use Only

Formula: C₂H₈ClN₃
MW: 109.557 g/mol
MDL Number: MFCD00012576
CAS Number: 21770-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR