Curcumin 98%

Supplier: Ambeed
Warning

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

A147405-1000G A147405-100G A147405-25G A147405-500G A147405-5G A147405-10G
77286-008EA 213 USD
77286-008 77286-010 77286-012 77286-014 77286-016 77567-040
Curcumin 98%
Curcumin

Precaución: For research use only.

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
MDL Number: MFCD00008365
CAS Number: 458-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR