5-Bromo-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol

A147342-100G A147342-10G A147342-500G A147342-5G A147342-25G
77285-994EA 27 USD
77285-994 77285-996 77285-998 77286-000 77299-576
5-Bromo-2-hydroxypyridine 98%
5-Bromo-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄BrNO
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR