4-(4-Ethylpiperazin-1-ylmethyl)phenylamine 95%

Supplier: Ambeed
Warning

A147237-10G A147237-1G A147237-5G
77285-984EA 302.82 USD
77285-984 77285-986 77285-988
4-(4-Ethylpiperazin-1-ylmethyl)phenylamine 95%
4-(4-Ethylpiperazin-1-ylmethyl)phenylamine

Precaución: for Research Use Only

Formula: C₁₃H₂₁N₃
MW: 219.3300 g/mol
MDL Number: MFCD05237186
CAS Number: 611225-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR