3,4-Dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Veratric acid

A147235-1000G A147235-100G A147235-25G A147235-500G
77247-154EA 132.87 USD
77247-154 77247-156 77317-214 77746-032
3,4-Dimethoxybenzoic acid 98%
3,4-Dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002500
CAS Number: 93-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR