4,5-Dibromo-2-phenylpyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A147182-1G A147182-250MG A147182-5G
77285-978EA 6.18 USD
77285-978 77285-980 77285-982
4,5-Dibromo-2-phenylpyridazin-3(2H)-one 97%
4,5-Dibromo-2-phenylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂N₂O
MW: 329.99 g/mol
MDL Number: MFCD00052907
CAS Number: 14305-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR