2'-Methylcytidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,3R,4R,5R)-3,4-dihydroxy-5-(hydroxymethyl)-3-methyltetrahydrofuran-2-yl)pyrimidin-2(1H)-one, NM107

A147052-100MG A147052-10MG A147052-1G A147052-1MG A147052-50MG A147052-5G A147052-5MG
77259-070EA 69.01 USD
77259-070 77259-072 77259-074 77259-076 77259-078 77259-080 77259-082
2'-Methylcytidine 98%
2'-Methylcytidine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₅
MW: 257.2400 g/mol
MDL Number: MFCD02682947
CAS Number: 20724-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR