Methyl 3-(hydroxymethyl)-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

A147033-1G A147033-5G
77235-236EA 51 USD
77235-236 77235-238
Methyl 3-(hydroxymethyl)-4-nitrobenzoate 97%
Methyl 3-(hydroxymethyl)-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD29762434
CAS Number: 201932-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR