2,2'-Bicinchoninic acid disodium salt 96%

Supplier: Ambeed
Warning

Synonyms: Sodium [2,2'-biquinoline]-4,4'-dicarboxylate, BCA disodium salt, Disodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-Biquinoline-4,4'-dicarboxylic acid disodium salt, Disodium 2,2'-Biquinoline-4,4'-dicarboxylate, Bicinchoninic acid disodium salt

A146947-10G A146947-1G A146947-1MG A146947-5G A146947-100G A146947-25G
77247-142EA 29 USD
77247-142 77247-144 77247-146 77247-148 77549-804 77567-008
2,2'-Bicinchoninic acid disodium salt 96%
2,2'-Bicinchoninic acid disodium salt

Precaución: For research use only.

Formula: C₂₀H₁₀N₂Na₂O₄
MDL Number: MFCD00037500
CAS Number: 979-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR