2,3-Difluoro-6-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A146940-10G A146940-1G A146940-250MG A146940-25G A146940-5G
77272-832EA 157.59 USD
77272-832 77272-834 77272-836 77272-838 77272-840
2,3-Difluoro-6-methoxyphenylboronic acid 98%
2,3-Difluoro-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
MDL Number: MFCD03095116
CAS Number: 957061-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR