1,3-Benzothiazole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Benzo[d]thiazole-6-carboxylic acid, Benzothiazole-6-carboxylic acid

A146923-1G A146923-250MG A146923-5G
77235-230EA 18.54 USD
77235-230 77235-232 77235-234
1,3-Benzothiazole-6-carboxylic acid 95%
1,3-Benzothiazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂S
MW: 179.2 g/mol
MDL Number: MFCD00111651
CAS Number: 3622-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR