3-Bromo-2-chloropyridine 98%

Supplier: Ambeed
Warning

A146888-100G A146888-10G A146888-25G A146888-500G A146888-5G
77285-964EA 55.62 USD
77285-964 77285-966 77285-968 77285-970 77285-972
3-Bromo-2-chloropyridine 98%
3-Bromo-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
MW: 192.44 g/mol
MDL Number: MFCD00234007
CAS Number: 52200-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR