2-Bromo-4-methylquinoline 98%

Supplier: Ambeed
Warning

A146730-1G A146730-250MG A146730-5G
77259-052EA 129.78 USD
77259-052 77259-054 77259-056
2-Bromo-4-methylquinoline 98%
2-Bromo-4-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
MDL Number: MFCD08705663
CAS Number: 64658-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR