3-(3-Nitrophenyl)propionic acid 98%

Supplier: Ambeed
Warning

A146716-100G A146716-10G A146716-250MG A146716-25G A146716-500G A146716-5G
77272-810EA 59 USD
77272-810 77272-812 77272-814 77272-816 77272-818 77272-820 77272-822
3-(3-Nitrophenyl)propionic acid 98%
3-(3-Nitrophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD01310835
CAS Number: 1664-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR