5,5'-(Anthracene-9,10-diyl)diisophthalic acid 98%

Supplier: Ambeed
Warning

A146567-100MG A146567-1G A146567-250MG A146567-5G
77259-032EA 28.84 USD
77259-032 77259-034 77259-036 77259-038
5,5'-(Anthracene-9,10-diyl)diisophthalic acid 98%
5,5'-(Anthracene-9,10-diyl)diisophthalic acid

Precaución: for Research Use Only

Formula: C₃₀H₁₈O₈
MW: 506.45 g/mol
MDL Number: MFCD31700771
CAS Number: 422269-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR