(5-Iodo-2-methylphenyl)methanol 97%

Supplier: Ambeed
Warning

A146453-10G A146453-1G A146453-250MG A146453-25G A146453-5G
77235-216EA 94.76 USD
77235-216 77235-218 77235-220 77235-222 77235-224
(5-Iodo-2-methylphenyl)methanol 97%
(5-Iodo-2-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉IO
MW: 248.061 g/mol
MDL Number: MFCD18397814
CAS Number: 1260242-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR