9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-[9-([1,1'-Biphenyl]-3-yl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9-(3-Biphenylyl)carbazole-3-boronic acid Pinacol ester

A146403-1G A146403-5G
77235-212EA 5 USD
77235-212 77235-214
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₈BNO₂
MW: 445.37 g/mol
MDL Number: MFCD28130384
CAS Number: 1533406-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR