4-Methylphthalic anhydride 98%

Supplier: Ambeed
Warning

A146319-100G A146319-25G
77285-948EA 118 USD
77285-948 77285-950
4-Methylphthalic anhydride 98%
4-Methylphthalic anhydride

Precaución: for Research Use Only

Formula: C₉H₆O₃
MDL Number: MFCD00041856
CAS Number: 19438-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR