3-Bromo-4,7-diazaindole 95%

Supplier: Ambeed
Warning

A146245-1G A146245-250MG A146245-5G
77272-804EA 131.84 USD
77272-804 77272-806 77272-808
3-Bromo-4,7-diazaindole 95%
3-Bromo-4,7-diazaindole

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MDL Number: MFCD09834812
CAS Number: 56015-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR