(3aS,8aS)-(2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine 98%

Supplier: Ambeed
Warning

A146234-100MG A146234-250MG A146234-1G
77247-124EA 30 USD
77247-124 77247-126 77746-026
(3aS,8aS)-(2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine 98%
(3aS,8aS)-(2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine

Precaución: for Research Use Only

Formula: C₃₃H₃₄NO₄P
CAS Number: 582300-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR