ONO-4059 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-6-amino-9-(1-(but-2-ynoyl)pyrrolidin-3-yl)-7-(4-phenoxyphenyl)-7,9-dihydro-8H-purin-8-one, Tirabrutinib

A146088-10MG A146088-25MG A146088-50MG A146088-5MG A146088-1MG
77235-194EA 56.65 USD
77235-194 77235-196 77235-198 77235-200 77615-166
ONO-4059 98+%
ONO-4059

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₆O₃
MW: 454.48 g/mol
MDL Number: MFCD28386296
CAS Number: 1351636-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR