4-Bromo-N-methoxy-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A146062-10G A146062-1G
77285-932EA 35 USD
77285-932 77285-934
4-Bromo-N-methoxy-N-methylbenzamide 98%
4-Bromo-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD02684303
CAS Number: 192436-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR