2-Amino-6-chloro-4-methylpyridine 97%

Supplier: Ambeed
Warning

A145959-10G A145959-1G A145959-250MG A145959-25G A145959-5G
77259-006EA 129 USD
77259-006 77259-008 77259-010 77259-012 77259-014
2-Amino-6-chloro-4-methylpyridine 97%
2-Amino-6-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD09839256
CAS Number: 51564-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR