3-Methyladenine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

A145872-100MG A145872-1MG A145872-250MG A145872-50MG
77272-792EA 25.75 USD
77272-792 77272-794 77272-796 77272-798
3-Methyladenine 98%
3-Methyladenine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00010531
CAS Number: 5142-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR