(1S)-1-(Diphenylphosphino)-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene 97%

Supplier: Ambeed
Warning

A145740-100MG A145740-250MG
77247-114EA 88 USD
77247-114 77746-018
(1S)-1-(Diphenylphosphino)-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene 97%
(1S)-1-(Diphenylphosphino)-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene

Precaución: for Research Use Only

Formula: C₃₆H₃₂FeP₂
MDL Number: MFCD30475629
CAS Number: 155941-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR