3-Chloro-4-methoxybenzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)-2-chloro-1-methoxybenzene

A145736-100MG A145736-10G A145736-1G A145736-250MG
77285-920EA 9.27 USD
77285-920 77285-922 77285-924 77285-926
3-Chloro-4-methoxybenzyl bromide 95%
3-Chloro-4-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
MDL Number: MFCD06858766
CAS Number: 320407-92-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR