3-(Trimethoxysilyl)propyl methacrylate 98% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
Stabilization:
100 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A145689-100G
A145689-25G
A145689-500G
77272-780EA
55.62
USD
77272-780
77272-782
77272-784
77272-786
3-(Trimethoxysilyl)propyl methacrylate 98% stabilised
3-(Trimethoxysilyl)propyl methacrylate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀O₅Si MW: 248.35 g/mol |
MDL Number:
MFCD00008593 CAS Number: 2530-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...