2-Oxoindoline-7-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A145679-100MG
A145679-250MG
A145679-50MG
77272-774EA
39
USD
77272-774
77272-776
77272-778
2-Oxoindoline-7-carboxylic acid 95%
2-Oxoindoline-7-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ |
MDL Number:
MFCD06208245 CAS Number: 25369-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...