(R)-1-(2-Chloro-6-fluorophenyl)ethanamine 97%

Supplier: Ambeed
Danger

A145674-1G A145674-250MG
77247-106EA 39 USD
77247-106 77247-108
(R)-1-(2-Chloro-6-fluorophenyl)ethanamine 97%
(R)-1-(2-Chloro-6-fluorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₉ClFN
MDL Number: MFCD06761974
CAS Number: 1100575-44-7
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR