4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Supplier: Ambeed
Warning

Synonyms: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

A145664-100G A145664-10G A145664-25G A145664-500G A145664-5G
77272-764EA 53.56 USD
77272-764 77272-766 77272-768 77272-770 77272-772
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆
MW: 323.35 g/mol
MDL Number: MFCD00065564
CAS Number: 7536-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR