2,6-Difluoro-4-pyridineboronic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2,6-Difluoropyridin-4-yl)boronic acid, 2,6-Difluoro-4-pyridylboronic acid
A145637-100MG
A145637-1G
A145637-250MG
77272-750EA
25
USD
77272-750
77272-752
77272-754
2,6-Difluoro-4-pyridineboronic acid 96%
2,6-Difluoro-4-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₅H₄BF₂NO₂ MW: 158.8986 g/mol |
MDL Number:
MFCD04114289 CAS Number: 401816-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...