Fmoc-D-Ala-OH.H2O 97%
Supplier: Ambeed
|
Warning
|
A145563-100G
A145563-10G
A145563-25G
A145563-500G
A145563-5G
77272-730EA
65
USD
77272-730
77272-732
77272-734
77272-736
77272-738
Fmoc-D-Ala-OH.H2O 97%
Fmoc-D-Ala-OH.H2O
Precaución: for Research Use Only
Formula:
C₁₈H₁₉NO₅ |
MDL Number:
MFCD00180722 CAS Number: 884880-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...