2-(1-Cyclohexenyl)cyclohexanone 85%

Supplier: Ambeed
Warning

Synonyms: [1,1'-Bi(cyclohexan)]-1'-en-2-one

A145509-100G A145509-5G
77235-188EA 60 USD
77235-188 77566-950
2-(1-Cyclohexenyl)cyclohexanone 85%
2-(1-Cyclohexenyl)cyclohexanone

Precaución: For research use only.

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
MDL Number: MFCD00013796
CAS Number: 1502-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR