4-(Chloromethyl)benzyl alcohol 97%

Supplier: Ambeed
Danger

Synonyms: p-hydroxymethylbenzyl chloride, (4-(Chloromethyl)phenyl)methanol, 4-(hydroxymethyl)benzyl chloride, Benzenemethanol,4-(chloromethyl)-

A145491-1G A145491-250MG A145491-5G A145491-10G A145491-25G
77285-914EA 11 USD
77285-914 77285-916 77285-918 77615-164 77748-014
4-(Chloromethyl)benzyl alcohol 97%
4-(Chloromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD03427011
CAS Number: 16473-35-1
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR